Anti-PPP1R13L Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34898-100EA 713 EUR
ABCAAB34898-100
Anti-PPP1R13L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPP1R13L (protein phosphatase 1, regulatory subunit 13 like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR