Anti-EGFR Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB34901-100EA 703 EUR
ABCAAB34901-100
Anti-EGFR Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR