Anti-HSP90AA1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34909-100EA 703 EUR
ABCAAB34909-100
Anti-HSP90AA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSP90AA1 (heat shock protein 90kDa alpha (cytosolic), class A member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR