Anti-Ganglioside asialo GM1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB34975-250EA 666 EUR
ABCAAB34975-250
Anti-Ganglioside asialo GM1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Ganglioside asialo GM1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR