Anti-beta subunit Cholera Toxin Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB34992-100EA 682 EUR
ABCAAB34992-100
Anti-beta subunit Cholera Toxin Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: beta subunit Cholera Toxin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR