Anti-Dexamethasone Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB35000-1EA 877 EUR
ABCAAB35000-1
Anti-Dexamethasone Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Dexamethasone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen