Anti-G protein alpha 12 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB35016-500EA 666 EUR
ABCAAB35016-500
Anti-G protein alpha 12 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: G protein alpha 12
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR