Anti-KLRK1 Mouse Monoclonal Antibody [clone: 1D11]

Lieferant: Abcam
ABCAAB35033-50EA 688 EUR
ABCAAB35033-50
Anti-KLRK1 Mouse Monoclonal Antibody [clone: 1D11]
Antikörper
Type: Primary
Antigen: KLRK1 (killer cell lectin-like receptor subfamily K, member 1)
Clonality: monoclonal
Clone: 1D11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR