Anti-KLRK1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 1D11]
Lieferant: Abcam
ABCAAB35035-50EA 876 EUR
ABCAAB35035-50
Anti-KLRK1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 1D11]
Antikörper
Type: Primary
Antigen: KLRK1 (killer cell lectin-like receptor subfamily K, member 1)
Clonality: monoclonal
Clone: 1D11
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human