Anti-WNK1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB35069-200EA 657 EUR
ABCAAB35069-200
Anti-WNK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: WNK1 (WNK lysine deficient protein kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR