Anti-GDF2 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB35088-500EA 703 EUR
ABCAAB35088-500
Anti-GDF2 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GDF2 (growth differentiation factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR