Anti-Salmonella Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB35156-1EA 672 EUR
ABCAAB35156-1
Anti-Salmonella Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Salmonella
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR