Anti-Staphylococcus aureus Rabbit Polyclonal Antibody (Biotin)

Lieferant: Abcam
ABCAAB35192-1EA 666 EUR
ABCAAB35192-1
Anti-Staphylococcus aureus Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Staphylococcus aureus
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR