Anti-E. coli pili Sheep polyclonal antibody

Lieferant: Abcam
ABCAAB35292-100EA 590 EUR
ABCAAB35292-100
Anti-E. coli pili Sheep polyclonal antibody
Type: Primary
Antigen: E. coli pili
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: E. coli
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR