Anti-NPY1R Sheep Polyclonal Antibody

Lieferant: Abcam
ABCAAB35336-500EA 895 EUR
ABCAAB35336-500
Anti-NPY1R Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NPY1R (neuropeptide Y receptor Y1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR