Anti-Pan Orexin Sheep Polyclonal Antibody

Lieferant: Abcam
ABCAAB35337-500EA 666 EUR
ABCAAB35337-500
Anti-Pan Orexin Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Pan Orexin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR