Anti-DCN Sheep polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB35378-1EA 666 EUR
ABCAAB35378-1
Anti-DCN Sheep polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DCN (decorin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR