Anti-Pancreatic Amylase Sheep Polyclonal Antibody

Lieferant: Abcam
ABCAAB35414-100EA 690 EUR
ABCAAB35414-100
Anti-Pancreatic Amylase Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Pancreatic Amylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR