Anti-CEA Mouse monoclonal antibody unconjugated [clone: CB30]

Lieferant: Abcam
ABCAAB35657-100EA 666 EUR
ABCAAB35657-100
Anti-CEA Mouse monoclonal antibody unconjugated [clone: CB30]
Antikörper
Type: Primary
Antigen: CEA (Carcino Embryonic Antigen)
Clonality: monoclonal
Clone: CB30
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR