Anti-SLC2A4 Mouse Monoclonal Antibody [clone: 1F8]

Lieferant: Abcam
ABCAAB35826-50EA 688 EUR
ABCAAB35826-50
Anti-SLC2A4 Mouse Monoclonal Antibody [clone: 1F8]
Antikörper
Type: Primary
Antigen: SLC2A4 (solute carrier family 2 (facilitated glucose transporter), member 4)
Clonality: monoclonal
Clone: 1F8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR