Anti-SLC2A5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB36057-50EA 882 EUR
ABCAAB36057-50
Anti-SLC2A5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC2A5 (solute carrier family 2 (facilitated glucose/fructose transporter), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR