Anti-COL1/2/3/4/5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB36064-50EA 861 EUR
ABCAAB36064-50
Anti-COL1/2/3/4/5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: COL1/2/3/4/5 (collagen, type I/II/III/IV/V)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR