Anti-C4d Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB36075-50EA 697 EUR
ABCAAB36075-50
Anti-C4d Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: C4d
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR