Anti-DLX4 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB36122-100EA 697 EUR
ABCAAB36122-100
Anti-DLX4 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DLX4 (distal-less homeobox 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR