Anti-RAB11FIP1 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB36124-100EA 657 EUR
ABCAAB36124-100
Anti-RAB11FIP1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAB11FIP1 (RAB11 family interacting protein 1 (class I))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR