Anti-SART3 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB36137-100EA 672 EUR
ABCAAB36137-100
Anti-SART3 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SART3 (squamous cell carcinoma antigen recognized by T cells 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR