Anti-GST3 / GST pi Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB36149-100EA 696 EUR
ABCAAB36149-100
Anti-GST3 / GST pi Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GST3 / GST pi
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR