Anti-NANOS1 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB36163-100EA 706 EUR
ABCAAB36163-100
Anti-NANOS1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NANOS1 (nanos homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR