Anti-VAMP7 Mouse Monoclonal Antibody [clone: 158.2]

Lieferant: Abcam
ABCAAB36195-50EA 910 EUR
ABCAAB36195-50
Anti-VAMP7 Mouse Monoclonal Antibody [clone: 158.2]
Antikörper
Type: Primary
Antigen: VAMP7 (vesicle-associated membrane protein 7)
Clonality: monoclonal
Clone: 158.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR