Anti-GCG Sheep polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB36215-100EA 666 EUR
ABCAAB36215-100
Anti-GCG Sheep polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GCG (glucagon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR