Anti-LYZ Mouse Monoclonal Antibody [clone: BGN/06/961]

Lieferant: Abcam
ABCAAB36362-100EA 784 EUR
ABCAAB36362-100
Anti-LYZ Mouse Monoclonal Antibody [clone: BGN/06/961]
Type: Primary
Antigen: LYZ (lysozyme)
Clonality: monoclonal
Clone: BGN/06/961
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR