Anti-Enterovirus 71 Mouse monoclonal antibody unconjugated [clone: 10F0]
Lieferant: Abcam
ABCAAB36367-50EA 876 EUR
ABCAAB36367-50
Anti-Enterovirus 71 Mouse monoclonal antibody unconjugated [clone: 10F0]
Antikörper
Type: Primary
Antigen: Enterovirus 71
Clonality: monoclonal
Clone: 10F0
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen