Anti-AKR1A1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB36376-100EA 666 EUR
ABCAAB36376-100
Anti-AKR1A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKR1A1 (aldo-keto reductase family 1, member A1 (aldehyde reductase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR