Anti-Factor D Mouse Monoclonal Antibody [clone: I8/1]

Lieferant: Abcam
ABCAAB36383-100EA 882 EUR
ABCAAB36383-100
Anti-Factor D Mouse Monoclonal Antibody [clone: I8/1]
Antikörper
Type: Primary
Antigen: Factor D
Clonality: monoclonal
Clone: I8/1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR