Anti-Natural Killer Cell Mouse Monoclonal Antibody [clone: ANK44]

Lieferant: Abcam
ABCAAB36388-500EA 666 EUR
ABCAAB36388-500
Anti-Natural Killer Cell Mouse Monoclonal Antibody [clone: ANK44]
Antikörper
Type: Primary
Antigen: Natural Killer Cell
Clonality: monoclonal
Clone: ANK44
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR