Anti-Protein C Mouse Monoclonal Antibody [clone: 6B2]

Lieferant: Abcam
ABCAAB36407-100EA 672 EUR
ABCAAB36407-100
Anti-Protein C Mouse Monoclonal Antibody [clone: 6B2]
Antikörper
Type: Primary
Antigen: Protein C
Clonality: monoclonal
Clone: 6B2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR