Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-181]

Lieferant: Abcam
ABCAAB36419-50EA 666 EUR
ABCAAB36419-50
Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-181]
Antikörper
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: monoclonal
Clone: MEM-181
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR