Anti-CD38 Mouse monoclonal antibody PE (Phycoerythrin) [clone: HIT2]

Lieferant: Abcam
ABCAAB36422-50EA 389 EUR
ABCAAB36422-50
Anti-CD38 Mouse monoclonal antibody PE (Phycoerythrin) [clone: HIT2]
Type: Primary
Antigen: CD38 (CD38 molecule)
Clonality: monoclonal
Clone: HIT2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR