Anti-VEGFA Mouse Monoclonal Antibody [clone: 6D3]

Lieferant: Abcam
ABCAAB36424-100EA 697 EUR
ABCAAB36424-100
Anti-VEGFA Mouse Monoclonal Antibody [clone: 6D3]
Antikörper
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone: 6D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR