Anti-CD52 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HI186]

Lieferant: Abcam
ABCAAB36443-50EA 666 EUR
ABCAAB36443-50
Anti-CD52 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HI186]
Antikörper
Type: Primary
Antigen: CD52 (CD52 molecule)
Clonality: monoclonal
Clone: HI186
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR