Anti-MAP2a + MAP2b Mouse Monoclonal Antibody [clone: MT-07]

Lieferant: Abcam
ABCAAB36447-100EA 706 EUR
ABCAAB36447-100
Anti-MAP2a + MAP2b Mouse Monoclonal Antibody [clone: MT-07]
Antikörper
Type: Primary
Antigen: MAP2a + MAP2b
Clonality: monoclonal
Clone: MT-07
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR