Anti-LAT2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB36452-100EA 672 EUR
ABCAAB36452-100
Anti-LAT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LAT2 (linker for activation of T cells family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR