Anti-TGF beta 2 Mouse Monoclonal Antibody
Lieferant: Abcam
ABCAAB36495-100EA 884 EUR
ABCAAB36495-100
Anti-TGF beta 2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TGF beta 2
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen