Anti-IGF1 Mouse Monoclonal Antibody

Lieferant: Abcam
ABCAAB36532-100EA 682 EUR
ABCAAB36532-100
Anti-IGF1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: IGF1 (insulin-like growth factor 1 (somatomedin C))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR