Anti-Reptin/TIP49B/RUVB2 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB36569-100EA 929 EUR
ABCAAB36569-100
Anti-Reptin/TIP49B/RUVB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Reptin/TIP49B/RUVB2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen