Anti-BCL10 Mouse monoclonal antibody unconjugated [clone: 4F8E8H8]

Lieferant: Abcam
ABCAAB36585-100EA 913 EUR
ABCAAB36585-100
Anti-BCL10 Mouse monoclonal antibody unconjugated [clone: 4F8E8H8]
Antikörper
Type: Primary
Antigen: BCL10 (B-cell CLL/lymphoma 10)
Clonality: monoclonal
Clone: 4F8E8H8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR