Anti-CD14 Mouse Monoclonal Antibody [clone: 5A3B11B5]

Lieferant: Abcam
ABCAAB36595-100EA 672 EUR
ABCAAB36595-100
Anti-CD14 Mouse Monoclonal Antibody [clone: 5A3B11B5]
Antikörper
Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: monoclonal
Clone: 5A3B11B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR