Anti-Lysosomal acid lipase Mouse Monoclonal Antibody [clone: 9G7F12]

Lieferant: Abcam
ABCAAB36597-100EA 688 EUR
ABCAAB36597-100
Anti-Lysosomal acid lipase Mouse Monoclonal Antibody [clone: 9G7F12]
Antikörper
Type: Primary
Antigen: Lysosomal acid lipase
Clonality: monoclonal
Clone: 9G7F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR