Anti-GCG Mouse monoclonal antibody unconjugated [clone: 1B7B4]

Lieferant: Abcam
ABCAAB36598-100EA 672 EUR
ABCAAB36598-100
Anti-GCG Mouse monoclonal antibody unconjugated [clone: 1B7B4]
Antikörper
Type: Primary
Antigen: GCG (glucagon)
Clonality: monoclonal
Clone: 1B7B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR