Anti-KSHV ORF45 Mouse monoclonal antibody [clone: 2D4A5]

Lieferant: Abcam
ABCAAB36618-100EA 603 EUR
ABCAAB36618-100
Anti-KSHV ORF45 Mouse monoclonal antibody [clone: 2D4A5]
Type: Primary
Antigen: KSHV ORF45
Clonality: monoclonal
Clone: 2D4A5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR